Order Entry
Export
ContactUsLinkComponent
 
Irritant
Decaethylene glycol monomethyl ether
Catalog # APOSBIPG1565-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Decaethylene glycol monomethyl ether
Catalog # APOSBIPG1565-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :472.57 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:27425-92-9
Formula:C₂₁H₄₄O₁₁
MDL Number:MFCD06797087