0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Hydroxy-PEG6-t-butyl ester
Hydroxy-PEG6-t-butyl ester
Hydroxy-PEG6-t-butyl ester
Irritant
Hydroxy-PEG6-t-butyl ester
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Hydroxy-PEG6-t-butyl ester
Supplier:
Apollo Scientific
About this item
CAS Number:
361189-64-2
:
Formula:
C₁₉H₃₈O₉
:
410.5 g/mol
:
GHS07
Storage Temperature:
Freezer