Order Entry
Export
ContactUsLinkComponent
Hydroxy-PEG4-t-butyl ester
Irritant
Hydroxy-PEG4-t-butyl ester
Catalog # APOSBIPG1450-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Hydroxy-PEG4-t-butyl ester
Catalog # APOSBIPG1450-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :332.39 g/mol
Density:1.04 g/cm³ (25 °C)
 :GHS07
Storage Temperature:Ambient
CAS Number:518044-32-1
Formula:C₁₅H₃₀O₇