Biotin-PEG4-amine
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: N-(14-Amino-3,6,9,12-tetraoxatetradecyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide, Amine-PEG4-Biotin, N-Biotinyl-3,6,9,12-tetraoxatetradecane-1,14-diamine, N-[2-[2-[2-[2-(2-Aminoethoxy)ethoxy]ethoxy]ethoxy]ethyl]biotinamide
- CAS Number:663171-32-2
- Formula:C₂₀H₃₈N₄O₆S
- MDL Number:MFCD26142992
- Molecular Weight:462.606 g/mol
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: N-(14-Amino-3,6,9,12-tetraoxatetradecyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide, Amine-PEG4-Biotin, N-Biotinyl-3,6,9,12-tetraoxatetradecane-1,14-diamine, N-[2-[2-[2-[2-(2-Aminoethoxy)ethoxy]ethoxy]ethoxy]ethyl]biotinamide
- CAS Number:663171-32-2
- Formula:C₂₀H₃₈N₄O₆S
- MDL Number:MFCD26142992
- Molecular Weight:462.606 g/mol
- Storage Temperature:Ambient



