Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
11-Azido-3,6,9-trioxaundecanol
11-Azido-3,6,9-trioxaundecanol
11-Azido-3,6,9-trioxaundecanol
Irritant
11-Azido-3,6,9-trioxaundecanol
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
11-Azido-3,6,9-trioxaundecanol
Supplier:
Apollo Scientific
About this item
CAS Number:
86770-67-4
:
Formula:
C₈H₁₇N₃O₄
MDL Number:
MFCD03701128
:
219.24 g/mol
:
GHS07
Storage Temperature:
Refrigerator