PD 98059
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Specific inhibitor of MAPK kinase
Synonyms: PD98059, 2'-Amino-3'-methoxyflavone, 2-(2-Amino-3-methoxyphenyl)chromone, PD 98059, 2-(2-Amino-3-methoxyphenyl)-4H-1-benzopyran-4-one, 2-(2-Amino-3-methoxyphenyl)-4H-chromen-4-one
- CAS Number:167869-21-8
- Formula:C₁₆H₁₃NO₃
- MDL Number:MFCD00671789
- Molecular Weight:267.284 g/mol
- Storage Temperature:Freezer
3 Options Available Below
Product Details & Documents
Specific inhibitor of MAPK kinase
Synonyms: PD98059, 2'-Amino-3'-methoxyflavone, 2-(2-Amino-3-methoxyphenyl)chromone, PD 98059, 2-(2-Amino-3-methoxyphenyl)-4H-1-benzopyran-4-one, 2-(2-Amino-3-methoxyphenyl)-4H-chromen-4-one
- CAS Number:167869-21-8
- Formula:C₁₆H₁₃NO₃
- MDL Number:MFCD00671789
- Molecular Weight:267.284 g/mol
- Storage Temperature:Freezer



