0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
1,2-O-Isopropylidene-alpha-D-xylofuranose
1,2-O-Isopropylidene-ɑ-D-xylofuranose
1,2-O-Isopropylidene-ɑ-D-xylofuranose
APOSBIO1700-5G
:
Apollo Scientific
:
1,2-O-Isopropylidene-ɑ-D-xylofuranose
APOSBIO1700-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
190.20 g/mol
:
112…114 °C/0.06mmHg
:
69…71 °C
:
GHS07
:
Refrigerator
:
20031-21-4
:
C₈H₁₄O₅
:
MFCD00063295