Order Entry
Export
Orders LinkContactUsLinkComponent
Basic blue 12

Basic blue 12

Irritant
Supplier:   Apollo Scientific
Basic blue 12
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents

Fluorecence reference standard.

Synonyms:  Nile Blue A chloride, 5-Amino-9-(diethylamino)benzo[a]phenoxazin-7-ium chloride
  • CAS Number:2381-85-3
  • Formula:C₂₀H₂₀ClN₃O
  • MDL Number:MFCD00011936
  • Molecular Weight:353.85 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 43,50 €
    43,50 €
    1 * 10 g
       43,50 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 72,40 €
    72,40 €
    1 * 25 g
       72,40 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 218,00 €
    218,00 €
    1 * 100 g
       218,00 €