



Malachite green oxalate
Catalog # APOSBIM2437-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:927.02 g/mol
Melting Pt:144…150 °C
Storage Temperature:Ambient
EINECS:219-441-7
UN:2811
ADR:6.1,III
:GHS05
:GHS06
:GHS08
:GHS09
CAS Number:2437-29-8
:18015-76-4
Formula:C₅₂H₅₄N₄O₁₂
MDL Number:MFCD00011766