Order Entry
Export
ContactUsLinkComponent
MOPS-Na ≥98%
MOPS-Na ≥98%
Catalog # APOSBIM0197-500G
Supplier:  Apollo Scientific
CAS Number:  
MOPS-Na ≥98%
Catalog # APOSBIM0197-500G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :231.25 g/mol
Storage Temperature:Ambient
CAS Number:71119-22-7
Formula:C₇H₁₄NNaO₄S
MDL Number:MFCD00064350