Order Entry
Export
Orders LinkContactUsLinkComponent
Linoleic acid

Linoleic acid

Irritant
Supplier:   Apollo Scientific
Linoleic acid
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid
  • CAS Number:60-33-3
  • Formula:C₁₈H₃₂O₂
  • MDL Number:MFCD00064241
  • Molecular Weight:280.45 g/mol
  • Boiling Pt:230 °C/15mmHg
  • Melting Pt:–12 °C
  • Density:0.902 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Freezer
  • EINECS:200-470-9
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 49,20 €
    49,20 €
    1 * 10 g
       49,20 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 79,60 €
    79,60 €
    1 * 25 g
       79,60 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 218,00 €
    218,00 €
    1 * 100 g
       218,00 €