Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D(+)-Raffinose pentahydrate
D-(+)-Raffinose pentahydrate
D-(+)-Raffinose pentahydrate
Irritant
D-(+)-Raffinose pentahydrate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
D-(+)-Raffinose pentahydrate
Supplier:
Apollo Scientific
About this item
CAS Number:
17629-30-0
:
Formula:
C₁₈H₃₂O₁₆·5H₂O
MDL Number:
MFCD00071590
:
594.52 g/mol
EINECS:
208-146-9
:
GHS07
Storage Temperature:
Ambient