Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L(+)-Arabinose
L(+)-Arabinose 98%
L(+)-Arabinose 98%
L(+)-Arabinose 98%
Supplier:
Apollo Scientific
Certificates
Expand image
L(+)-Arabinose 98%
Supplier:
Apollo Scientific
Certificates
About this item
CAS Number:
5328-37-0
:
87-72-9
Formula:
C₅H₁₀O₅
MDL Number:
MFCD00067709
:
150.13 g/mol
Melting Pt:
160…163 °C
EINECS:
201-767-6
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
Documentation