5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:69-78-3
:
Formula:C₁₄H₈N₂O₈S₂
MDL Number:MFCD00007140
:396.36 g/mol
Melting Pt:233…243 °C
EINECS:200-714-4
:GHS07
Storage Temperature:Ambient