17-DMAG
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 17-Dimethylaminoethylamino-17-demethoxygeldanamycin
- CAS Number:467214-20-6
- Formula:C₃₂H₄₈N₄O₈
- MDL Number:MFCD08457919
- Molecular Weight:616.8 g/mol
- Storage Temperature:Freezer
4 Options Available Below
Product Details & Documents
Synonyms: 17-Dimethylaminoethylamino-17-demethoxygeldanamycin
- CAS Number:467214-20-6
- Formula:C₃₂H₄₈N₄O₈
- MDL Number:MFCD08457919
- Molecular Weight:616.8 g/mol
- Storage Temperature:Freezer
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



