Methyl-Β-L-arabinopyranoside 98% min
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: (2S,3R,4S,5S)-2-Methoxytetrahydro-2H-pyran-3,4,5-triol
CAS Number:1825-00-9
:
Formula:C₆H₁₂O₅
MDL Number:MFCD00047547
:164.17 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: