D-Cycloserine
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: D-(+)-Cycloserine, Cycloserine, D(+)-Cycloserine, (R)-4-Aminoisoxazolidin-3-one, D-4-Aminoisoxazolidin-3-one, (R)-(+)-Cycloserine
CAS Number:68-41-7
:
Formula:C₃H₆N₂O₂
MDL Number:MFCD00005353
Molecular Weight:102.09 g/mol
Melting Pt:145…147 °C (dec.)
EINECS:200-688-4
:GHS07
Storage Temperature:Freezer
Specifications
- Environmentally Preferable: