Order Entry
Export
Orders LinkContactUsLinkComponent
Bisacrylamide ≥98%

Bisacrylamide ≥98%

Irritant
Supplier:   Apollo Scientific
Bisacrylamide ≥98%
Supplier:  Apollo Scientific
Certificates
Bisacrylamide ≥98%

3 Options Available Below

Product Details & Documents
Synonyms:  Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS
  • CAS Number:110-26-9
  • Formula:(H₂C=CHCONH)₂CH₂
  • MDL Number:MFCD00008625
  • Molecular Weight:154.17 g/mol
  • Melting Pt:>300 °C
  • Melting Pt:>300 °C
  • Storage Temperature:Refrigerator
  • EINECS:203-750-9
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 71,00 €
    71,00 €
    1 * 100 g
       71,00 €

  • Bottle
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 138,00 €
    138,00 €
    1 * 250 g
       138,00 €

  • Bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 283,00 €
    283,00 €
    1 * 1 kg
       283,00 €