Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
PIPES
PIPES ≥99.5%, Ultrapure
PIPES ≥99.5%, Ultrapure
Irritant
PIPES ≥99.5%, Ultrapure
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
PIPES ≥99.5%, Ultrapure
Supplier:
Apollo Scientific
About this item
CAS Number:
5625-37-6
:
Formula:
C₈H₁₈N₂O₆S₂
MDL Number:
MFCD00006159
:
302.37 g/mol
Melting Pt:
>300 °C
EINECS:
227-057-6
:
GHS07
Storage Temperature:
Ambient