
Bromocresol green sodium salt
Catalog # APOSBIB1187-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
Melting Pt:230 °C (dec.)
:720 g/mol
EINECS:263-657-4
:GHS07
Storage Temperature:Ambient
CAS Number:62625-32-5
:67763-24-0
Formula:C₂₁H₁₃Br₄NaO₅S
MDL Number:MFCD00148898