
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid ≥99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:10191-18-1
:
Formula:C₆H₁₅NO₅S
MDL Number:MFCD00007533
Melting Pt:153…157 °C
:213.25 g/mol
EINECS:233-465-5
:GHS07
Storage Temperature:Ambient