17-AAG
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Hsp90 inhibitor.
Synonyms: 17-(Allylamino)-17-demethoxygeldanamycin, Tanespimycin, (4E,6Z,8S,9S,10E,12S,13R,14S,16R)-19-(Allylamino)-13-hydroxy-8,14-dimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate
- CAS Number:75747-14-7
- Formula:C₃₁H₄₃N₃O₈
- MDL Number:MFCD04973892
- Molecular Weight:585.7 g/mol
- Storage Temperature:Freezer
3 Options Available Below
Product Details & Documents
Hsp90 inhibitor.
Synonyms: 17-(Allylamino)-17-demethoxygeldanamycin, Tanespimycin, (4E,6Z,8S,9S,10E,12S,13R,14S,16R)-19-(Allylamino)-13-hydroxy-8,14-dimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate
- CAS Number:75747-14-7
- Formula:C₃₁H₄₃N₃O₈
- MDL Number:MFCD04973892
- Molecular Weight:585.7 g/mol
- Storage Temperature:Freezer



