Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Aminoactinomycin D (7-AAD)
7-Aminoactinomycin D (7-AAD)
7-Aminoactinomycin D (7-AAD)
Toxic
Health
7-Aminoactinomycin D (7-AAD)
Supplier:
Apollo Scientific
Certificates
Expand image
Toxic
Health
7-Aminoactinomycin D (7-AAD)
Supplier:
Apollo Scientific
About this item
CAS Number:
7240-37-1
:
Formula:
C₆₂H₈₇N₁₃O₁₆
MDL Number:
MFCD00077295
:
1270.43 g/mol
UN:
3462
ADR:
6.1,II
:
GHS06
:
GHS08
Storage Temperature:
Freezer