Order Entry
Export
Orders LinkContactUsLinkComponent
Taurine ≥99%

Taurine ≥99%

Irritant
Supplier:   Apollo Scientific
Taurine ≥99%
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents
  • Water (by Karl Fischer) <1.0%
Synonyms:  2-Aminoethanesulphonic acid
  • CAS Number:107-35-7
  • Formula:C₂H₇NO₃S
  • MDL Number:MFCD00008197
  • Molecular Weight:125.15 g/mol
  • Melting Pt:>300 °C
  • Melting Pt:>300 °C
  • Storage Temperature:Ambient
  • EINECS:203-483-8

Documents

Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 21,80 €
    21,80 €
    1 * 100 g
       21,80 €

    Specifications:
    More
    Specifications
    Cat. No.
    APOSBIA0603-100G
  • Bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 24,60 €
    24,60 €
    1 * 500 g
       24,60 €

    Specifications:
    More
    Specifications
    Cat. No.
    APOSBIA0603-500G
  • Bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 36,20 €
    36,20 €
    1 * 1 kg
       36,20 €

    Specifications:
    More
    Specifications
    Cat. No.
    APOSBIA0603-1KG