Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L-Alanine-tert-butyl ester hydrochloride
L-Alanine-tert-butyl ester hydrochloride
L-Alanine-tert-butyl ester hydrochloride
Irritant
L-Alanine-tert-butyl ester hydrochloride
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
L-Alanine-tert-butyl ester hydrochloride
Supplier:
Apollo Scientific
About this item
CAS Number:
13404-22-3
:
Formula:
C₇H₁₅NO₂
MDL Number:
MFCD00035524
:
181.66 g/mol
EINECS:
236-497-8
:
GHS07
Storage Temperature:
Refrigerator