Amphotericin B
Supplier: Apollo Scientific
Jump to:
About this item
A polyene antifungal antibiotic produced by Streptomyces nodosus.
Synonyms: (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
- CAS Number:1397-89-3
- Formula:C₄₇H₇₃NO₁₇
- MDL Number:MFCD00877763
- Molecular Weight:924.08 g/mol
- Storage Temperature:Refrigerator
3 Options Available Below
Product Details & Documents
A polyene antifungal antibiotic produced by Streptomyces nodosus.
Synonyms: (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
- CAS Number:1397-89-3
- Formula:C₄₇H₇₃NO₁₇
- MDL Number:MFCD00877763
- Molecular Weight:924.08 g/mol
- Storage Temperature:Refrigerator



