Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dichlorobenzyl mercaptan
2,4-Dichlorobenzyl mercaptan 98%
2,4-Dichlorobenzyl mercaptan 98%
2,4-Dichlorobenzyl mercaptan 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2,4-Dichlorobenzyl mercaptan 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
59293-67-3
:
Formula:
C₇H₆Cl₂S
MDL Number:
MFCD00039650
:
193.10 g/mol
Storage Temperature:
Ambient