Bis-trispropane 99+%
Supplier: Apollo Scientific
Jump to:
About this item
Biological buffer - useful range 6.3-9.5
- Assay (by titration) 99+%
- Water (by Karl Fischer) < 1.0%
Synonyms: 1,3-Bis(tris(hydroxymethyl)methylamino)propane, 1,3-Bis[tris(hydroxymethyl)amino]propane, 2,2'-(Propane-1,3-diylbis(azanediyl))bis(2-(hydroxymethyl)propane-1,3-diol), BIS-TRIS propane, 2,2'-(Propane-1,3-diyldiimino)bis[2-(hydroxymethyl)propane-1,3-diol]
- CAS Number:64431-96-5
- Formula:C₁₁H₂₆N₂O₆
- MDL Number:MFCD00004689
- Molecular Weight:282.34 g/mol
- Melting Pt:164…165 °C
- Storage Temperature:Ambient
- EINECS:264-899-3
3 Options Available Below
Product Details & Documents
Biological buffer - useful range 6.3-9.5
- Assay (by titration) 99+%
- Water (by Karl Fischer) < 1.0%
Synonyms: 1,3-Bis(tris(hydroxymethyl)methylamino)propane, 1,3-Bis[tris(hydroxymethyl)amino]propane, 2,2'-(Propane-1,3-diylbis(azanediyl))bis(2-(hydroxymethyl)propane-1,3-diol), BIS-TRIS propane, 2,2'-(Propane-1,3-diyldiimino)bis[2-(hydroxymethyl)propane-1,3-diol]
- CAS Number:64431-96-5
- Formula:C₁₁H₂₆N₂O₆
- MDL Number:MFCD00004689
- Molecular Weight:282.34 g/mol
- Melting Pt:164…165 °C
- Storage Temperature:Ambient
- EINECS:264-899-3



