Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Phleomycin
Phleomycin
Phleomycin
:
Apollo Scientific
Phleomycin
:
Apollo Scientific
:
11006-33-0
:
:
MFCD00131846
:
GHS07
:
Freezer