Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Difluoro-4-iodo-2-methylsulphanylbenzene
1,3-Difluoro-4-iodo-2-methylsulphanylbenzene 97%
1,3-Difluoro-4-iodo-2-methylsulphanylbenzene 97%
1,3-Difluoro-4-iodo-2-methylsulphanylbenzene 97%
Supplier:
Apollo Scientific
Certificates
Expand image
1,3-Difluoro-4-iodo-2-methylsulphanylbenzene 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
2484889-05-4
:
Formula:
C₇H₅F₂IS
MDL Number:
MFCD32707289
:
286.081 g/mol
Storage Temperature:
Refrigerator