Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 2-chloro-3,4-difluorobenzoate
Ethyl 2-chloro-3,4-difluorobenzoate 95%
Ethyl 2-chloro-3,4-difluorobenzoate 95%
Ethyl 2-chloro-3,4-difluorobenzoate 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Ethyl 2-chloro-3,4-difluorobenzoate 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1261541-73-4
:
Formula:
C₉H₇ClF₂O₂
MDL Number:
MFCD18416422
:
220.6 g/mol
Storage Temperature:
Refrigerator