Order Entry
Export
ContactUsLinkComponent
4,5-Difluorobenzene-1,2-diol 97%
4,5-Difluorobenzene-1,2-diol 97%
Catalog # APOSPC9889-250MG
Supplier:  Apollo Scientific
CAS Number:  
4,5-Difluorobenzene-1,2-diol 97%
Catalog # APOSPC9889-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :146.092 g/mol
Storage Temperature:Ambient
CAS Number:147300-08-1
Formula:C₆H₄F₂O₂
MDL Number:MFCD08458183