
1H-Tridecafluoro-3,3-dimethylhex-1-yne
Catalog # APOSPC9873-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:344.073 g/mol
Boiling Pt:89 °C
Density:1.56 g/cm³ (25 °C)
:GHS07
Storage Temperature:Refrigerator
CAS Number:261503-44-0
Formula:C₈HF₁₃
MDL Number:MFCD00155807