
2-(2,4-Difluorophenyl)thiazole-4-carboxamide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175276-97-8
:
Formula:C₁₀H₆F₂N₂OS
MDL Number:MFCD00277199
:240.229 g/mol
Melting Pt:239…241 °C
:GHS07
Storage Temperature:Refrigerator