Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,4-Difluoro-6-azaspiro[2.5]octane hydrochloride
4,4-Difluoro-6-azaspiro[2.5]octane hydrochloride
4,4-Difluoro-6-azaspiro[2.5]octane hydrochloride
4,4-Difluoro-6-azaspiro[2.5]octane hydrochloride
Supplier:
Apollo Scientific
Certificates
Expand image
4,4-Difluoro-6-azaspiro[2.5]octane hydrochloride
Supplier:
Apollo Scientific
About this item
CAS Number:
1282532-00-6
:
Formula:
C₇H₁₁F₂N·ClH
MDL Number:
MFCD20669267
:
183.627 g/mol
Storage Temperature:
Ambient