Order Entry
Export
ContactUsLinkComponent
 
2-fluorobenzene-1,3-dicarboxylic acid 95%
Catalog # APOSPC910889-1G
Supplier:  Apollo Scientific
CAS Number:  
2-fluorobenzene-1,3-dicarboxylic acid 95%
Catalog # APOSPC910889-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :184.121 g/mol
Storage Temperature:Ambient
CAS Number:1583-65-9
Formula:C₈H₅FO₄
MDL Number:MFCD00042505