Order Entry
Export
ContactUsLinkComponent
3,5-Dichloro-4-fluorophenol 96%
3,5-Dichloro-4-fluorophenol 96%
Catalog # APOSPC910271-1G
Supplier:  Apollo Scientific
CAS Number:  
3,5-Dichloro-4-fluorophenol 96%
Catalog # APOSPC910271-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :180.992 g/mol
Storage Temperature:Ambient
CAS Number:2995-04-2
Formula:C₆H₃Cl₂FO
MDL Number:MFCD19690558