Order Entry
Export
ContactUsLinkComponent
4-Ethoxy-2,5-difluorophenol 95%
4-Ethoxy-2,5-difluorophenol 95%
Catalog # APOSPC910266-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Ethoxy-2,5-difluorophenol 95%
Catalog # APOSPC910266-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :174.145 g/mol
Storage Temperature:Ambient
CAS Number:612092-32-7
Formula:C₈H₈F₂O₂
MDL Number:MFCD24728303