Order Entry
Export
ContactUsLinkComponent
3-Chloro-2,4-difluorophenol 95%
3-Chloro-2,4-difluorophenol 95%
Catalog # APOSPC910261-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Chloro-2,4-difluorophenol 95%
Catalog # APOSPC910261-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :164.537 g/mol
Storage Temperature:Ambient
CAS Number:2268-00-0
Formula:C₆H₃ClF₂O
MDL Number:MFCD18415629