Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Difluoro-5-methoxyphenol
3,4-Difluoro-5-methoxyphenol 95%
3,4-Difluoro-5-methoxyphenol 95%
APOSPC910259-1G
:
Apollo Scientific
:
3,4-Difluoro-5-methoxyphenol 95%
APOSPC910259-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
160.118 g/mol
:
Ambient
:
1394915-77-5
:
C₇H₆F₂O₂
:
MFCD22414498