Order Entry
Export
ContactUsLinkComponent
3,5-Difluoro-2-methoxyphenol 96%
3,5-Difluoro-2-methoxyphenol 96%
Catalog # APOSPC910240-1G
Supplier:  Apollo Scientific
CAS Number:  
3,5-Difluoro-2-methoxyphenol 96%
Catalog # APOSPC910240-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :160.118 g/mol
Storage Temperature:Ambient
CAS Number:152434-94-1
Formula:C₇H₆F₂O₂
MDL Number:MFCD12404933