Order Entry
Export
ContactUsLinkComponent
2-Chloro-3,4-difluoroanisole 95%
2-Chloro-3,4-difluoroanisole 95%
Catalog # APOSPC910119-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-3,4-difluoroanisole 95%
Catalog # APOSPC910119-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :178.564 g/mol
Storage Temperature:Ambient
CAS Number:1261769-08-7
Formula:C₇H₅ClF₂O
MDL Number:MFCD18415541