Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-2,3-difluorophenol
4-Chloro-2,3-difluorophenol 95%
4-Chloro-2,3-difluorophenol 95%
APOSPC910092-1G
:
Apollo Scientific
:
4-Chloro-2,3-difluorophenol 95%
APOSPC910092-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
164.537 g/mol
:
Ambient
:
1261634-63-2
:
C₆H₃ClF₂O
:
MFCD18415633