Order Entry
Export
ContactUsLinkComponent
4-Bromo-2,3-difluoropyridine 95%
4-Bromo-2,3-difluoropyridine 95%
Catalog # APOSPC909991-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-2,3-difluoropyridine 95%
Catalog # APOSPC909991-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.977 g/mol
Storage Temperature:Ambient
CAS Number:1227597-53-6
Formula:C₅H₂BrF₂N
MDL Number:MFCD16609910