Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Fluoroimidazo[1,2-a]pyridine hydrochloride
5-Fluoroimidazo[1,2-a]pyridine hydrochloride 95%
5-Fluoroimidazo[1,2-a]pyridine hydrochloride 95%
5-Fluoroimidazo[1,2-a]pyridine hydrochloride 95%
APOSPC908467-1G
:
Apollo Scientific
:
5-Fluoroimidazo[1,2-a]pyridine hydrochloride 95%
APOSPC908467-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
172.587 g/mol
:
Ambient
:
198896-14-9
:
C₇H₅FN₂·ClH
:
MFCD08705707