Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Dichloro-3-fluorobenzoic acid
2,5-Dichloro-3-fluorobenzoic acid 95%
2,5-Dichloro-3-fluorobenzoic acid 95%
2,5-Dichloro-3-fluorobenzoic acid 95%
APOSPC907983-25G
:
Apollo Scientific
:
2,5-Dichloro-3-fluorobenzoic acid 95%
APOSPC907983-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
209.002 g/mol
:
Ambient
:
501008-42-0
:
C₇H₃Cl₂FO₂
:
MFCD13250174