Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chloro-2,4-difluorobromobenzene
5-Chloro-2,4-difluorobromobenzene
5-Chloro-2,4-difluorobromobenzene
Irritant
5-Chloro-2,4-difluorobromobenzene
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Chloro-2,4-difluorobromobenzene
Supplier:
Apollo Scientific
About this item
CAS Number:
914636-89-8
:
Formula:
C₆H₂BrClF₂
MDL Number:
MFCD04112557
:
227.434 g/mol
:
GHS07
Storage Temperature:
Ambient