Order Entry
Export
ContactUsLinkComponent
2-(2,3-Difluorophenyl)acetaldehyde 95%
2-(2,3-Difluorophenyl)acetaldehyde 95%
Catalog # APOSPC907200-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(2,3-Difluorophenyl)acetaldehyde 95%
Catalog # APOSPC907200-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :156.129 g/mol
Storage Temperature:Ambient
CAS Number:866342-07-6
Formula:C₈H₆F₂O
MDL Number:MFCD11553568