Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 2,3-difluoroisonicotinate
Methyl 2,3-difluoroisonicotinate ≥98%
Methyl 2,3-difluoroisonicotinate ≥98%
Methyl 2,3-difluoroisonicotinate ≥98%
Supplier:
Apollo Scientific
Certificates
Expand image
Methyl 2,3-difluoroisonicotinate ≥98%
Supplier:
Apollo Scientific
About this item
CAS Number:
1353102-03-0
:
Formula:
C₇H₅F₂NO₂
MDL Number:
MFCD21090433
:
173.117 g/mol
Storage Temperature:
Ambient