Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-1,5-difluoro-3-nitrobenzene
2-Bromo-1,5-difluoro-3-nitrobenzene 95%
2-Bromo-1,5-difluoro-3-nitrobenzene 95%
2-Bromo-1,5-difluoro-3-nitrobenzene 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Bromo-1,5-difluoro-3-nitrobenzene 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
877161-74-5
:
Formula:
C₆H₂BrF₂NO₂
MDL Number:
MFCD04112512
:
237.986 g/mol
Storage Temperature:
Ambient